Anti-DMRT1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2752]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8075-97H-FITCEA 0 DKK
USBID8075-97H-FITC
Anti-DMRT1 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2752]
Type: Primary
Antigen: DMRT1 (doublesex and mab-3 related transcription factor 1)
Clonality: monoclonal
Clone: 10B2752
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR