Anti-DMPO Nitrone Adduct Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 12F1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8075-94CEA 4090 DKK
USBID8075-94C
Anti-DMPO Nitrone Adduct Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 12F1]
Antistoffer
Type: Primary
Antigen: DMPO Nitrone Adduct
Clonality: monoclonal
Clone: 12F1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR