Anti-DMPK Mouse Monoclonal Antibody [clone: 12K82]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8075-83EEA 4320 DKK
USBID8075-83E
Anti-DMPK Mouse Monoclonal Antibody [clone: 12K82]
Antistoffer
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: DMPK (dystrophia myotonica-protein kinase)
Clonality: monoclonal
Clone: 12K82
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR