Anti-DMPK Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8075-83AEA 3980 DKK
USBID8075-83A
Anti-DMPK Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 0.1-1 ?g/ml; Western Blot: 1-3?g/ml

Type: Primary
Antigen: DMPK (dystrophia myotonica-protein kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR