Anti-DMP1 Mouse Monoclonal Antibody (Biotin) [clone: 11D61]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8075-51BEA 5320 DKK
USBID8075-51B
Anti-DMP1 Mouse Monoclonal Antibody (Biotin) [clone: 11D61]
Antistoffer
Recommended dilution: ELISA: 0.5-2?g/ml

Type: Primary
Antigen: DMP1 (dentin matrix acidic phosphoprotein 1)
Clonality: monoclonal
Clone: 11D61
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR