Anti-Dentin Matrix Protein 1 Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8075-51EA 4930 DKK
USBID8075-51
Anti-Dentin Matrix Protein 1 Sheep polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Dentin Matrix Protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR