Anti-DMC1 Mouse Monoclonal Antibody [clone: 8J1306]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8075-01EA 4410 DKK
USBID8075-01
Anti-DMC1 Mouse Monoclonal Antibody [clone: 8J1306]
Antistoffer
Type: Primary
Antigen: DMC1 (DNA meiotic recombinase 1)
Clonality: monoclonal
Clone: 8J1306
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR