Anti-DMC Rat Monoclonal Antibody [clone: 9H112]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8074-90CEA 4320 DKK
USBID8074-90C
Anti-DMC Rat Monoclonal Antibody [clone: 9H112]
Antistoffer
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 2?g/ml

Type: Primary
Antigen: DMC
Clonality: monoclonal
Clone: 9H112
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR