Anti-DMC Mouse Monoclonal Antibody [clone: 7H252]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8074-90BEA 4320 DKK
USBID8074-90B
Anti-DMC Mouse Monoclonal Antibody [clone: 7H252]
Antistoffer
Recommended dilution: Flow Cytometry: 25 ?g/ml; Immunochemistry: 10 ?g/ml; Western Blot: 1-2?g/ml

Type: Primary
Antigen: DMC
Clonality: monoclonal
Clone: 7H252
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR