Anti-DISC1 Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8066-83FEA 4780 DKK
USBID8066-83F
Anti-DISC1 Sheep polyclonal antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 5-15 ?g/ml

Type: Primary
Antigen: DISC1 (disrupted in schizophrenia 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR