Anti-DISC1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8066-83AEA 3310 DKK
USBID8066-83A
Anti-DISC1 Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: DISC1 (disrupted in schizophrenia 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR