Anti-Diphtheria Toxin Mouse monoclonal antibody [clone: 9B55]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID8065-48BEA 3410 DKK
USBID8065-48B
Anti-Diphtheria Toxin Mouse monoclonal antibody [clone: 9B55]
Antistoffer
Type: Primary
Antigen: Diphtheria Toxin
Clonality: monoclonal
Clone: 9B55
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu