Anti-Diphtheria Toxin Goat Polyclonal Antibody (FITC (Fluorescein))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8065-47EEA 2970 DKK
USBID8065-47E
Anti-Diphtheria Toxin Goat Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Recommended dilution: Immunofluorescence: 1:5-1:25

Type: Primary
Antigen: Diphtheria Toxin
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR