Anti-Diphtheria Toxin Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8065-47DEA 3330 DKK
USBID8065-47D
Anti-Diphtheria Toxin Goat polyclonal antibody
Antistoffer
Type: Primary
Antigen: Diphtheria Toxin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR