Anti-Diphtheria Toxin Goat Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8065-47BEA 2970 DKK
USBID8065-47B
Anti-Diphtheria Toxin Goat Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Diphtheria Toxin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR