Anti-Diphtheria Toxin Mouse monoclonal antibody [clone: 10B2749]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8065-42CEA 4870 DKK
USBID8065-42C
Anti-Diphtheria Toxin Mouse monoclonal antibody [clone: 10B2749]
Antistoffer
Type: Primary
Antigen: Diphtheria Toxin
Clonality: monoclonal
Clone: 10B2749
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR