Anti-DPP10 Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8065-33EA 4500 DKK
USBID8065-33
Anti-DPP10 Sheep Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DPP10 (dipeptidyl-peptidase 10 (non-functional))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR