Anti-DPP9 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8065-31KEA 4730 DKK
USBID8065-31K
Anti-DPP9 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: DPP9 (dipeptidyl-peptidase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR