Anti-Dipeptidyl Peptidase II Rat monoclonal antibody [clone: 8B15]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8065-31BEA 4320 DKK
USBID8065-31B
Anti-Dipeptidyl Peptidase II Rat monoclonal antibody [clone: 8B15]
Antistoffer
Type: Primary
Antigen: Dipeptidyl Peptidase II
Clonality: monoclonal
Clone: 8B15
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR