Anti-Dendroaspis Natriuretic Peptide Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8060-13EA 5090 DKK
USBID8060-13
Anti-Dendroaspis Natriuretic Peptide Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:3200

Type: Primary
Antigen: Dendroaspis Natriuretic Peptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR