Anti-DNP Mouse Monoclonal Antibody [clone: 9L190]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8060-10AEA 3590 DKK
USBID8060-10A
Anti-DNP Mouse Monoclonal Antibody [clone: 9L190]
Antistoffer
Type: Primary
Antigen: DNP (Dinitrophenol)
Clonality: monoclonal
Clone: 9L190
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR