Anti-DNP Mouse Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8060-02BEA 5040 DKK
USBID8060-02B
Anti-DNP Mouse Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000-1:10000; Western Blot: 1:200-1:2000

Type: Primary
Antigen: DNP (Dinitrophenol)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR