Anti-DNP Rat Monoclonal Antibody [clone: 10B2748]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8060-01FEA 6730 DKK
USBID8060-01F
Anti-DNP Rat Monoclonal Antibody [clone: 10B2748]
Antistoffer
  • Recommended dilution: ELISA: 1. 25 μg/ml

Type: Primary
Antigen: DNP (Dinitrophenol)
Clonality: monoclonal
Clone: 10B2748
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgE
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR