Anti-Dihydrotestosterone Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8036-03EA 5050 DKK
USBID8036-03
Anti-Dihydrotestosterone Sheep polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 2.5 ?g/ml

Type: Primary
Antigen: Dihydrotestosterone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR