Anti-DHFR Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8034-20CEA 3740 DKK
USBID8034-20C
Anti-DHFR Rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: DHFR (dihydrofolate reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR