Anti-Digoxin Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8030-22AEA 2510 DKK
USBID8030-22A
Anti-Digoxin Goat polyclonal antibody
Antistoffer
Type: Primary
Antigen: Digoxin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR