Anti-Digoxin Mouse monoclonal antibody [clone: 8.F.80]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8030-17-500EA 4590 DKK
USBID8030-17-500 USBID8030-17-1
Anti-Digoxin Mouse monoclonal antibody [clone: 8.F.80]
Antistoffer
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone: 8.F.80
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR