Anti-Digoxin Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8030-11CEA 2770 DKK
USBID8030-11C
Anti-Digoxin Sheep polyclonal antibody
Antistoffer
Recommended dilution: Radio Immunoassay: 1:6000

Type: Primary
Antigen: Digoxin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR