Anti-Digoxigenin Sheep polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID8029-50JEA 6540 DKK
USBID8029-50J
Anti-Digoxigenin Sheep polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 0. 625 ?g/ml

Type: Tag
Antigen: Digoxigenin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR