Anti-DLK2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID7800-40-HRPEA 4620 DKK
USBID7800-40-HRP
Anti-DLK2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: DLK2 (delta-like 2 homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR