Anti-DHX9 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2746]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID6509-85D-FITCEA 0 DKK
USBID6509-85D-FITC
Anti-DHX9 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2746]
Type: Primary
Antigen: DHX9 (DEAH (Asp-Glu-Ala-His) box helicase 9)
Clonality: monoclonal
Clone: 10B2746
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR