Anti-DHPR, alpha 2 Mouse monoclonal antibody [clone: 6D357]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID6500-10AEA 4280 DKK
USBID6500-10A
Anti-DHPR, alpha 2 Mouse monoclonal antibody [clone: 6D357]
Antistoffer
Recommended dilution: Immunochemistry: 1:250-500; Western Blot: 1:500

Type: Primary
Antigen: DHPR, alpha 2
Clonality: monoclonal
Clone: 6D357
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Guinea pig, Human, Mouse, Rabbit, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR