Anti-DNAJC14 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2743]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID4015-08F1-APCEA 0 DKK
USBID4015-08F1-APC
Anti-DNAJC14 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2743]
Type: Primary
Antigen: DNAJC14 (DnaJ (Hsp40) homolog, subfamily C, member 14)
Clonality: monoclonal
Clone: 10B2743
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR