Anti-DNAJC14 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2741]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID4015-08A5-APEA 0 DKK
USBID4015-08A5-AP
Anti-DNAJC14 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2741]
Type: Primary
Antigen: DNAJC14 (DnaJ (Hsp40) homolog, subfamily C, member 14)
Clonality: monoclonal
Clone: 10B2741
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR