Anti-DNAJA3 Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID4015-07UEA 4730 DKK
USBID4015-07U
Anti-DNAJA3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: DNAJA3 (DnaJ (Hsp40) homolog, subfamily A, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse