Anti-DNA Methyltransferase 3B Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-73BEA 3240 DKK
USBID3931-73B
Anti-DNA Methyltransferase 3B Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: DNA Methyltransferase 3B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR