Anti-DNMT3A Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-70SEA 3180 DKK
USBID3931-70S
Anti-DNMT3A Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1-3?g/ml

Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR