Anti-DNMT3A Mouse Monoclonal Antibody (Biotin) [clone: 11E170]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-70QEA 4050 DKK
USBID3931-70Q
Anti-DNMT3A Mouse Monoclonal Antibody (Biotin) [clone: 11E170]
Antistoffer
Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: monoclonal
Clone: 1.1E+171
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR