Anti-DNMT1 Chicken Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3931-63MEA 5180 DKK
USBID3931-63M
Anti-DNMT1 Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 2.5 ?g/ml; Western Blot: 1:2000
Type: Primary
Antigen: DNMT1 (DNA (cytosine-5-)-methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human