Anti-DNMT1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-63HEA 3700 DKK
USBID3931-63H
Anti-DNMT1 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500

Type: Primary
Antigen: DNMT1 (DNA (cytosine-5-)-methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR