Anti-DNA Methyltransferase 1 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-63BEA 4120 DKK
USBID3931-63B
Anti-DNA Methyltransferase 1 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-1. 5 ?g/ml

Type: Primary
Antigen: DNA Methyltransferase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR