Anti-TRDMT1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3931-62D-APCEA 6360 DKK
USBID3931-62D-APC
Anti-TRDMT1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse