Anti-DNA Methyltransferase 1, Control Peptide, Human Mouse Monoclonal Antibody [clone: 10F697]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3931-61SEA 4090 DKK
USBID3931-61S
Anti-DNA Methyltransferase 1, Control Peptide, Human Mouse Monoclonal Antibody [clone: 10F697]
Antistoffer
Recommended dilution: Western Blot:2-4?g/ml; Immunochemistry:1-2?g/ml I
Type: Primary
Antigen: DNA Methyltransferase 1, Control Peptide, Human
Clonality: Monoclonal
Clone: 10F697
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Zebrafish