Anti-DNA Methyltransferase 1 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-61JEA 3630 DKK
USBID3931-61J
Anti-DNA Methyltransferase 1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DNA Methyltransferase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR