Anti-DNA Methyltransferase 1 Mouse Monoclonal Antibody [clone: 6D354]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-61A3EA 4020 DKK
USBID3931-61A3
Anti-DNA Methyltransferase 1 Mouse Monoclonal Antibody [clone: 6D354]
Antistoffer
Type: Primary
Antigen: DNA Methyltransferase 1
Clonality: monoclonal
Clone: 6D354
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR