Anti-DNA Methyltransferase 1 Rabbit Monoclonal Antibody [clone: 10E19]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3931-614EA 4120 DKK
USBID3931-614
Anti-DNA Methyltransferase 1 Rabbit Monoclonal Antibody [clone: 10E19]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: DNA Methyltransferase 1
Clonality: monoclonal
Clone: 1E+20
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR