Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 10G12]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3925-02AEA 5250 DKK
USBID3925-02A
Anti-DNA Ligase 1 Mouse Monoclonal Antibody [clone: 10G12]
Antistoffer
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: DNA Ligase 1
Clonality: monoclonal
Clone: 10G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human