Anti-DNA/RNA Oxidative Damage Mouse Monoclonal Antibody [clone: 10F696]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3916-100EA 4540 DKK
USBID3916-100
Anti-DNA/RNA Oxidative Damage Mouse Monoclonal Antibody [clone: 10F696]
Antistoffer
Type: Primary
Antigen: DNA/RNA Oxidative Damage
Clonality: monoclonal
Clone: 10F696
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR