Anti-DNA, ds Mouse Monoclonal Antibody [clone: AE-2]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBID3878-13BEA 4030 DKK
USBID3878-13B
Anti-DNA, ds Mouse Monoclonal Antibody [clone: AE-2]
Antistoffer
Type: Primary
Antigen: DNA, ds
Clonality: monoclonal
Clone: AE-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human