Anti-DNA Mouse Monoclonal Antibody [clone: 10B2731]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBID3875-03EA 3150 DKK
USBID3875-03
Anti-DNA Mouse Monoclonal Antibody [clone: 10B2731]
Antistoffer
Recommended dilution: Flow Cytometry:1:10;

Type: Primary
Antigen: DNA
Clonality: monoclonal
Clone: 10B2731
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR